The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007997.1 | NP_032023.1 | 1 - 2 | 167 | 165 |
GenBank mRNA | AK134068.1 | - | 1 - 2 | 158 | 165 |
BC071268.1 | - | 1 - 2 | 167 | 165 | |
D49920.1 | - | 1 - 2 | 167 | 165 |