Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinf2
Entrez Gene ID
18816
Gene Name
serine (or cysteine) peptidase inhibitor, clade F, member 2
Gene Aliases
AI747498, Pli, Serpimf2
Chromosome Location
Chr.11: 75431736 - 75439501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008878.2 NP_032904.1 9 - 10 1109 85
GenBank mRNA AK077563.1 - 9 - 10 1111 85
BC026756.1 - 9 - 10 1083 85
GQ896353.1 - 8 - 9 1060 85
GQ896354.1 - 8 - 9 1060 85
Z36774.1 - 9 - 10 1096 85

View Full Product Details