Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smarcal1
Entrez Gene ID
54380
Gene Name
SWI/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1
Gene Aliases
6030401P21Rik, AI851716, Mharp
Chromosome Location
Chr.1: 72536687 - 72636790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018817.2 NP_061287.2 13 - 14 2366 69
GenBank mRNA AK043129.1 - 2 - 3 181 69
AK077878.1 - 13 - 14 2344 69
AK083488.1 - 13 - 14 2344 69
AK146767.1 - 13 - 14 2362 69
AK154020.1 - 13 - 14 2349 69
AK161570.1 - 12 - 13 3658 69
BC029078.1 - 13 - 14 2301 69

View Full Product Details