Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd28
Entrez Gene ID
12487
Gene Name
CD28 antigen
Gene Aliases
-
Chromosome Location
Chr.1: 60746388 - 60773359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007642.4 NP_031668.3 3 - 4 610 98
GenBank mRNA AK030812.1 - 3 - 4 532 98
AK088021.1 - 3 - 4 544 98
BC064058.1 - 3 - 4 524 98
M74361.1 - 1 - 2 59 98

View Full Product Details