Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgfbrap1
Entrez Gene ID
73122
Gene Name
transforming growth factor, beta receptor associated protein 1
Gene Aliases
3110018K12Rik, AU024090, AU042859, AW742551, Trap1
Chromosome Location
Chr.1: 43047265 - 43098622 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013025.2 NP_001013043.1 5 - 6 1602 96
GenBank mRNA AK141742.1 - 4 - 5 1158 96
BC072608.1 - 5 - 6 1602 96
BC138866.1 - 5 - 6 1516 96
BC138867.1 - 5 - 6 1516 96

View Full Product Details