The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103177.2 | NP_001096647.1 | 9 - 10 | 1212 | 96 |
NM_001103178.2 | NP_001096648.1 | 6 - 7 | 634 | 96 | |
NM_001290813.1 | NP_001277742.1 | 9 - 10 | 1198 | 96 | |
NM_001290815.1 | NP_001277744.1 | 9 - 10 | 1212 | 96 | |
NM_001290816.1 | NP_001277745.1 | 9 - 10 | 1212 | 96 | |
NM_178688.3 | NP_848803.3 | 11 - 12 | 1330 | 96 | |
GenBank mRNA | AF404774.1 | - | 11 - 12 | 1330 | 96 |
AF404775.1 | - | 9 - 10 | 972 | 96 | |
AF404776.1 | - | 6 - 7 | 594 | 96 | |
AK079331.1 | - | 5 - 6 | 452 | 96 | |
AK122196.1 | - | 9 - 10 | 1212 | 96 | |
AK143907.1 | - | 9 - 10 | 982 | 96 | |
AK171860.1 | - | - | 1109 | 96 | |
BC141083.1 | - | 9 - 10 | 1192 | 96 |