Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aamp
Entrez Gene ID
227290
Gene Name
angio-associated migratory protein
Gene Aliases
AU040907, Aamp-rs
Chromosome Location
Chr.1: 74279840 - 74284738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190444.1 NP_001177373.1 3 - 4 512 66
NM_146110.3 NP_666222.2 3 - 4 509 66
GenBank mRNA AK146470.1 - 3 - 4 452 66
AK151820.1 - 3 - 4 421 66
AK162750.1 - 3 - 4 437 66
AK167621.1 - 3 - 4 433 66
AK180202.1 - 2 - 3 153 66
AK200115.1 - 2 - 3 153 66
AK212494.1 - 2 - 3 153 66
BC031795.1 - 2 - 3 201 66
BC080766.1 - 1 - 2 920 66

View Full Product Details