Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pms1
Entrez Gene ID
227099
Gene Name
postmeiotic segregation increased 1 (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.1: 53189187 - 53297026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153556.1 NP_705784.1 8 - 9 1091 149
GenBank mRNA AK140689.1 - 9 - 10 1156 149
BC028939.1 - 8 - 9 1091 149

View Full Product Details