Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nop58
Entrez Gene ID
55989
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
MSSP, Nol5, SIK, nop5
Chromosome Location
Chr.1: 59684951 - 59711510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018868.2 NP_061356.2 4 - 5 479 108
GenBank mRNA AF053232.1 - 4 - 5 384 108
AK044216.1 - - 476 108
AK133959.1 - 4 - 5 477 108
AK134286.1 - 4 - 5 480 108
BC076604.1 - 4 - 5 480 108
BC085135.1 - 4 - 5 449 108

View Full Product Details