The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030013.2 | NP_084289.1 | 10 - 11 | 1095 | 63 |
GenBank mRNA | AK053388.1 | - | 10 - 11 | 1098 | 63 |
BC049147.1 | - | 9 - 10 | 912 | 63 |