Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Boll
Entrez Gene ID
75388
Gene Name
bol, boule-like (Drosophila)
Gene Aliases
4930554P13Rik, 4930597B14Rik
Chromosome Location
Chr.1: 55239385 - 55364058 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113367.1 NP_001106838.1 3 - 4 465 136
NM_029267.3 NP_083543.2 3 - 4 410 136
GenBank mRNA AF272859.1 - 3 - 4 273 136
AK016125.1 - 3 - 4 411 136
AK016403.1 - 3 - 4 465 136

View Full Product Details