Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Katnal2
Entrez Gene ID
71206
Gene Name
katanin p60 subunit A-like 2
Gene Aliases
3110023G01Rik, 4933439B08Rik
Chromosome Location
Chr.18: 76977147 - 77048225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027721.2 NP_081997.1 10 - 11 969 63
GenBank mRNA AK014074.1 - 11 - 12 1080 63
AK017114.1 - 10 - 11 970 63
BC056963.1 - 10 - 11 916 63

View Full Product Details