Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptchd2
Entrez Gene ID
242748
Gene Name
patched domain containing 2
Gene Aliases
G630052C06Rik, RNDEu-2
Chromosome Location
Chr.4: 148236433 - 148288161 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083342.1 NP_001076811.1 17 - 18 3484 70
GenBank mRNA AB235901.1 - 16 - 17 3238 70
AK078196.1 - 7 - 8 983 70
AK090326.1 - 17 - 18 3507 70
AK173152.1 - 16 - 17 2927 70
BC070441.1 - 17 - 18 3358 70
BC072569.1 - 17 - 18 3383 70

View Full Product Details