Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf8
Entrez Gene ID
320595
Gene Name
PHD finger protein 8
Gene Aliases
9830141C09Rik, mKIAA1111
Chromosome Location
Chr.X: 151520612 - 151633859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113354.1 NP_001106825.1 19 - 20 3389 90
NM_177201.5 NP_796175.1 18 - 19 3086 90
GenBank mRNA AK036609.1 - 18 - 19 2460 90
AK122447.1 - 18 - 19 2480 90
BC138899.1 - 18 - 19 2355 90

View Full Product Details