Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xrcc5
Entrez Gene ID
22596
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 5
Gene Aliases
AI314015, Ku80, Ku86
Chromosome Location
Chr.1: 72307417 - 72394953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009533.2 NP_033559.2 11 - 12 1374 68
GenBank mRNA AK089944.1 - 11 - 12 1371 68
AK165470.1 - 11 - 12 1342 68
AK167312.1 - 11 - 12 1263 68
AK168913.1 - 11 - 12 1263 68
AK169838.1 - 11 - 12 1263 68
BC029218.1 - 11 - 12 1246 68
BC051660.1 - 11 - 12 1314 68

View Full Product Details