Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pias2
Entrez Gene ID
17344
Gene Name
protein inhibitor of activated STAT 2
Gene Aliases
6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2
Chromosome Location
Chr.18: 77065208 - 77155710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164167.1 NP_001157639.1 10 - 11 1526 66
NM_001164168.1 NP_001157640.1 10 - 11 1542 66
NM_001164169.1 NP_001157641.1 10 - 11 1526 66
NM_001164170.1 NP_001157642.1 10 - 11 1542 66
NM_008602.4 NP_032628.3 10 - 11 1526 66
GenBank mRNA AF039567.1 - 9 - 10 971 66
AK014871.1 - 10 - 11 1517 66
AK029716.1 - 10 - 11 1527 66
AK078097.1 - 5 - 6 594 66
AK086653.1 - 10 - 11 1458 66
AK149142.1 - 11 - 12 1565 66
BC005596.1 - 10 - 11 1403 66
BC034711.1 - 10 - 11 1532 66

View Full Product Details