The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164167.1 | NP_001157639.1 | 2 - 3 | 690 | 118 |
NM_001164168.1 | NP_001157640.1 | 2 - 3 | 706 | 118 | |
NM_001164169.1 | NP_001157641.1 | 2 - 3 | 690 | 118 | |
NM_001164170.1 | NP_001157642.1 | 2 - 3 | 706 | 118 | |
NM_008602.4 | NP_032628.3 | 2 - 3 | 690 | 118 | |
GenBank mRNA | AF039567.1 | - | 1 - 2 | 135 | 118 |
AK014871.1 | - | 2 - 3 | 681 | 118 | |
AK029716.1 | - | 2 - 3 | 691 | 118 | |
AK078813.1 | - | 2 - 3 | 660 | 118 | |
AK086653.1 | - | 2 - 3 | 622 | 118 | |
AK149142.1 | - | 2 - 3 | 653 | 119 | |
BC005596.1 | - | 2 - 3 | 567 | 118 | |
BC034711.1 | - | 2 - 3 | 696 | 118 |