The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178749.3 | NP_848864.1 | 12 - 13 | 1339 | 115 |
GenBank mRNA | AK036266.1 | - | 12 - 13 | 1340 | 115 |
AK042599.1 | - | 12 - 13 | 1453 | 115 | |
AK044474.1 | - | 12 - 13 | 1349 | 115 | |
BC055002.1 | - | - | 1269 | 115 |