The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022996.1 | NP_075372.1 | 7 - 8 | 809 | 71 |
GenBank mRNA | AF220209.1 | - | 7 - 8 | 847 | 71 |
BC020359.1 | - | 7 - 8 | 796 | 71 | |
BC026372.1 | - | 7 - 8 | 809 | 71 |