The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033226.4 | NP_001028398.1 | 4 - 5 | 752 | 71 |
NM_001285895.1 | NP_001272824.1 | 5 - 6 | 716 | 71 | |
NM_001285896.1 | NP_001272825.1 | 5 - 6 | 729 | 71 | |
NM_001285897.1 | NP_001272826.1 | 5 - 6 | 971 | 71 | |
NM_001285898.1 | NP_001272827.1 | 5 - 6 | 981 | 71 | |
GenBank mRNA | AK077094.1 | - | 5 - 6 | 821 | 71 |
AK139540.1 | - | 5 - 6 | 731 | 71 | |
AK163350.1 | - | 4 - 5 | 753 | 71 | |
BC044740.2 | - | 5 - 6 | 981 | 71 | |
JN255799.1 | - | 5 - 6 | 716 | 71 |