Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc27a6
Entrez Gene ID
225579
Gene Name
solute carrier family 27 (fatty acid transporter), member 6
Gene Aliases
4732438L20Rik, FACVL2, FATP6, VLCS-H1
Chromosome Location
Chr.18: 58556240 - 58612869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081072.1 NP_001074541.1 3 - 4 1073 70
GenBank mRNA AK028699.1 - 3 - 4 1075 70
BC115435.1 - 3 - 4 1056 70

View Full Product Details