Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grp
Entrez Gene ID
225642
Gene Name
gastrin releasing peptide
Gene Aliases
BLP
Chromosome Location
Chr.18: 65872748 - 65886599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175012.3 NP_778177.1 1 - 2 273 87
GenBank mRNA AK133508.1 - 1 - 2 265 87
AK139345.1 - 1 - 2 268 87
BC024515.1 - 1 - 2 258 87

View Full Product Details