Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyrm5
Entrez Gene ID
67636
Gene Name
LYR motif containing 5
Gene Aliases
4930469P12Rik, AL022899, Ghiso
Chromosome Location
Chr.6: 145211134 - 145216542 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF412299.1 - 2 - 3 336 92

View Full Product Details