Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snapc3
- Entrez Gene ID
-
77634
- Gene Name
- small nuclear RNA activating complex, polypeptide 3
- Gene Aliases
-
1810020H02Rik, 4930558A07Rik, 5031401C21Rik, AI414457, E030018J20Rik
- Chromosome Location
-
Chr.4: 83417701 - 83467658 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019863.2 |
- |
11 - 12 |
2416 |
110 |
View Full Product Details