Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hspa12a
- Entrez Gene ID
-
73442
- Gene Name
- heat shock protein 12A
- Gene Aliases
-
1700063D12Rik, AI118035, AI840429, AW048913, AW556406, D5Mgi40, mKIAA0417
- Chromosome Location
-
Chr.19: 58795750 - 58943545 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076683.1 |
- |
2 - 3 |
266 |
66 |
View Full Product Details