Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem88b
Entrez Gene ID
320587
Gene Name
transmembrane protein 88B
Gene Aliases
A230069A22Rik
Chromosome Location
Chr.4: 155781591 - 155785874 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033394.3 NP_001028566.2 1 - 2 533 118
GenBank mRNA AK035998.1 - 1 - 2 533 118
AK158459.1 - 1 - 2 534 118
BC147318.1 - 1 - 2 488 118
BC147319.1 - 1 - 2 488 118

View Full Product Details