Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vwc2l
Entrez Gene ID
320460
Gene Name
von Willebrand factor C domain-containing protein 2-like
Gene Aliases
A830006F12Rik, Brl
Chromosome Location
Chr.1: 70725221 - 70886514 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177164.3 NP_796138.2 2 - 3 1194 108
GenBank mRNA AB374230.1 - 1 - 2 390 108
AK080585.1 - 4 - 5 643 108
BC094541.1 - 2 - 3 1194 108
EF552208.1 - 1 - 2 458 108

View Full Product Details