The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178241.4 | NP_839972.1 | 2 - 2 | 1064 | 60 |
GenBank mRNA | AY227797.1 | - | 1 - 1 | 1016 | 60 |
AY390263.1 | - | 2 - 2 | 1064 | 60 | |
AY749637.1 | - | 1 - 1 | 1390 | 60 |