The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195338.1 | NP_001182267.1 | 2 - 3 | 589 | 113 |
NM_001195348.1 | NP_001182277.1 | 2 - 3 | 328 | 113 | |
GenBank mRNA | AK009096.1 | - | 2 - 3 | 184 | 113 |
AK010048.1 | - | 2 - 3 | 329 | 113 | |
AK011697.1 | - | 2 - 3 | 134 | 113 | |
AK011849.1 | - | 2 - 3 | 134 | 113 | |
AK016185.1 | - | 2 - 3 | 199 | 113 | |
BC030472.1 | - | 1 - 2 | 136 | 113 | |
BC037041.1 | - | 1 - 2 | 121 | 113 | |
BC040803.1 | - | 2 - 3 | 151 | 113 | |
BC054437.1 | - | 2 - 3 | 143 | 113 | |
BC062276.1 | - | 2 - 3 | 190 | 113 | |
BC094636.1 | - | 1 - 2 | 119 | 113 |