Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema6c
Entrez Gene ID
20360
Gene Name
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C
Gene Aliases
Semay, mKIAA1869
Chromosome Location
Chr.3: 95160420 - 95174050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272024.1 NP_001258953.1 7 - 8 840 97
NM_011351.2 NP_035481.1 4 - 5 408 97
GenBank mRNA AB013729.1 - 4 - 5 373 97
AF363972.1 - 4 - 5 441 97
AF363973.1 - 4 - 5 441 97
BC059007.1 - 7 - 8 755 97

View Full Product Details