Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam45a
Entrez Gene ID
67894
Gene Name
family with sequence similarity 45, member A
Gene Aliases
1810055E12Rik
Chromosome Location
Chr.19: 60811441 - 60836235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167829.1 NP_001161301.1 4 - 5 527 146
NM_026437.3 NP_080713.2 4 - 5 527 146
GenBank mRNA AK004643.1 - 3 - 4 447 146
AK007867.1 - 4 - 5 487 146
AK145769.1 - 4 - 5 493 146
AK168474.1 - 3 - 4 441 146
BC013514.1 - 4 - 5 486 146

View Full Product Details