Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam45a
Entrez Gene ID
67894
Gene Name
family with sequence similarity 45, member A
Gene Aliases
1810055E12Rik
Chromosome Location
Chr.19: 60811441 - 60836235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167829.1 NP_001161301.1 7 - 8 833 109
NM_026437.3 NP_080713.2 7 - 8 854 109
GenBank mRNA AK004643.1 - 6 - 7 774 109
AK007867.1 - 7 - 8 814 109
AK145769.1 - 7 - 8 820 109
AK168474.1 - 6 - 7 768 109
BC013514.1 - 7 - 8 813 109

View Full Product Details