Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fam60a
- Entrez Gene ID
-
56306
- Gene Name
- family with sequence similarity 60, member A
- Gene Aliases
-
Ppcs1, Pptcs1, Tera
- Chromosome Location
-
Chr.6: 148921045 - 148946432 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK046519.1 |
- |
1 - 2 |
45 |
82 |
View Full Product Details