Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spats2l
Entrez Gene ID
67198
Gene Name
spermatogenesis associated, serine-rich 2-like
Gene Aliases
2810022L02Rik, A230104H11Rik, AA987173, AI480531, AI842453, AW413586
Chromosome Location
Chr.1: 57774162 - 57948398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164566.1 NP_001158038.1 1 - 2 109 82
GenBank mRNA BC014764.1 - 1 - 2 109 82
BY069430.1 - 1 - 2 65 82

View Full Product Details