Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rftn2
Entrez Gene ID
74013
Gene Name
raftlin family member 2
Gene Aliases
2700010E02Rik, 3222401M22Rik
Chromosome Location
Chr.1: 55170159 - 55227065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK012229.2 - 3 - 4 1146 123

View Full Product Details