Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ambp
Entrez Gene ID
11699
Gene Name
alpha 1 microglobulin/bikunin
Gene Aliases
AI194774, ASPI, HI-30, Intin4, Itil, UTI
Chromosome Location
Chr.4: 63143279 - 63154142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007443.3 NP_031469.1 5 - 6 635 96
GenBank mRNA BC021660.1 - 5 - 6 654 96
D28812.1 - 5 - 6 636 96
X68680.1 - 5 - 6 597 96

View Full Product Details