Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Espn
Entrez Gene ID
56226
Gene Name
espin
Gene Aliases
je
Chromosome Location
Chr.4: 152120331 - 152152420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207688.2 NP_997571.1 3 - 4 541 89
NM_207691.2 NP_997574.2 3 - 4 541 89
GenBank mRNA AF540942.1 - 3 - 4 541 89
AF540943.1 - 3 - 4 541 89

View Full Product Details