Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Espn
Entrez Gene ID
56226
Gene Name
espin
Gene Aliases
je
Chromosome Location
Chr.4: 152120331 - 152152420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207687.2 NP_997570.1 12 - 13 2110 85
NM_207688.2 NP_997571.1 7 - 8 1139 85
NM_207689.2 NP_997572.1 6 - 7 896 85
NM_207690.2 NP_997573.1 5 - 6 869 85
NM_207691.2 NP_997574.2 6 - 7 1112 85
GenBank mRNA AF239886.1 - 12 - 13 2110 85
AF540942.1 - 6 - 7 1112 85
AF540943.1 - 7 - 8 1139 85
AF540944.1 - 5 - 6 869 85
AF540945.1 - 6 - 7 896 85
AK145160.1 - 4 - 5 647 85
AK146701.1 - 4 - 5 614 85
AK165010.1 - 3 - 4 1619 85
AY587570.1 - 3 - 4 389 85
AY587571.1 - 2 - 3 479 85

View Full Product Details