Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inadl
Entrez Gene ID
12695
Gene Name
InaD-like (Drosophila)
Gene Aliases
Cipp, Patj
Chromosome Location
Chr.4: 98395534 - 98719603 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172696.2 NP_766284.2 25 - 26 3645 82
GenBank mRNA AK050545.1 - 21 - 22 2621 82
AK077268.1 - 12 - 13 1927 82
AK147866.1 - 25 - 26 3594 82
BC082787.1 - 14 - 15 2038 82
BC141404.1 - 25 - 26 3621 82

View Full Product Details