Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inadl
Entrez Gene ID
12695
Gene Name
InaD-like (Drosophila)
Gene Aliases
Cipp, Patj
Chromosome Location
Chr.4: 98395534 - 98719603 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172696.2 NP_766284.2 27 - 28 3822 70
GenBank mRNA AK077268.1 - 14 - 15 2104 70
AK147866.1 - 27 - 28 3771 70
BC082787.1 - 16 - 17 2215 70
BC141404.1 - 27 - 28 3798 70

View Full Product Details