The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028925.1 | NP_083201.1 | 10 - 11 | 1177 | 76 |
GenBank mRNA | AK016716.1 | - | 10 - 11 | 1204 | 76 |
AK177320.1 | - | 4 - 5 | 258 | 76 | |
BC147485.1 | - | 10 - 11 | 1177 | 76 | |
BC151028.1 | - | 10 - 11 | 1177 | 76 |