Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnn2
Entrez Gene ID
140492
Gene Name
potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2
Gene Aliases
KCa2.2, SK2, SKCA2, fri
Chromosome Location
Chr.18: 45268764 - 45685887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080465.2 NP_536713.1 6 - 7 1358 80
GenBank mRNA AF357240.1 - 6 - 7 1358 80
AF533008.1 - 6 - 7 1358 80
AK033158.1 - 9 - 10 2862 80
AK050390.1 - 6 - 7 1614 80
AK149082.1 - 6 - 7 2046 80
AY123778.1 - 6 - 7 1433 80
BC125475.1 - 6 - 7 1384 80
BC137656.1 - 6 - 7 1384 80
EU478850.1 - 5 - 6 938 80

View Full Product Details