Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tas1r1
Entrez Gene ID
110326
Gene Name
taste receptor, type 1, member 1
Gene Aliases
Gpr70, T1r1, TR1
Chromosome Location
Chr.4: 152027914 - 152038490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031867.2 NP_114073.1 2 - 3 601 71
GenBank mRNA AF301161.1 - 2 - 3 497 71
AF301162.1 - 2 - 3 497 71
AF337040.1 - 2 - 3 679 71
AK153579.1 - 2 - 3 551 71
AY032622.1 - 2 - 3 497 71
BC070448.1 - 2 - 3 601 71

View Full Product Details