Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmd3
Entrez Gene ID
242506
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, 9430066I12Rik, EPB41L4O, P410
Chromosome Location
Chr.4: 74013603 - 74202214 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163732.1 NP_001157204.1 6 - 7 768 135
NM_172869.4 NP_766457.1 6 - 7 768 135
GenBank mRNA AK034956.1 - 6 - 7 743 135
AK048561.1 - 6 - 7 711 135
AK079349.1 - 6 - 7 769 135
BC037727.1 - 6 - 7 939 135
BC112419.1 - 6 - 7 676 135
BC113795.1 - 6 - 7 676 135
BC118964.2 - 6 - 7 678 135

View Full Product Details