Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gys2
Entrez Gene ID
232493
Gene Name
glycogen synthase 2
Gene Aliases
BC021322, LGS
Chromosome Location
Chr.6: 142422612 - 142473124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145572.2 NP_663547.2 10 - 11 1621 161
GenBank mRNA AK138992.1 - 10 - 11 1329 161
AK149391.1 - 10 - 11 1345 161
BC021322.1 - 10 - 11 1482 161
BC158081.1 - 10 - 11 1482 161

View Full Product Details