The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271587.1 | NP_001258516.1 | 16 - 17 | 2004 | 78 |
NM_001271588.1 | NP_001258517.1 | 16 - 17 | 1935 | 78 | |
NM_001271589.1 | NP_001258518.1 | 9 - 10 | 1005 | 78 | |
NM_001271595.1 | NP_001258524.1 | 16 - 17 | 2009 | 78 | |
NM_007945.3 | NP_031971.2 | 17 - 18 | 2010 | 78 | |
GenBank mRNA | AK136246.1 | - | 7 - 8 | 776 | 78 |
AK147877.1 | - | 17 - 18 | 1938 | 78 | |
AK149683.1 | - | 16 - 17 | 1798 | 78 | |
AK155022.1 | - | 16 - 17 | 1770 | 78 | |
AK165684.1 | - | 9 - 10 | 1008 | 78 | |
AK166156.1 | - | 17 - 18 | 1845 | 78 | |
BC016890.1 | - | 16 - 17 | 2021 | 78 | |
L21671.1 | - | 16 - 17 | 1925 | 78 |