The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271587.1 | NP_001258516.1 | 20 - 21 | 2675 | 76 |
NM_001271588.1 | NP_001258517.1 | 20 - 21 | 2606 | 76 | |
NM_001271589.1 | NP_001258518.1 | 13 - 14 | 1676 | 76 | |
NM_001271595.1 | NP_001258524.1 | 20 - 21 | 2680 | 76 | |
NM_007945.3 | NP_031971.2 | 21 - 22 | 2681 | 76 | |
GenBank mRNA | AK136246.1 | - | 11 - 12 | 1448 | 76 |
AK149683.1 | - | 20 - 21 | 2469 | 76 | |
AK155022.1 | - | 20 - 21 | 2441 | 76 | |
AK165684.1 | - | 13 - 14 | 1679 | 76 | |
AK166156.1 | - | 21 - 22 | 2516 | 76 | |
BC016890.1 | - | 20 - 21 | 2692 | 76 | |
L21671.1 | - | 20 - 21 | 2596 | 76 |