Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zc3h4
- Entrez Gene ID
-
330474
- Gene Name
- zinc finger CCCH-type containing 4
- Gene Aliases
-
Bwq1, Gm768, Kiaa1064-hp, mKIAA1064
- Chromosome Location
-
Chr.7: 16399572 - 16437698 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198631.2 |
NP_941033.2 |
5 - 6 |
821 |
94 |
View Full Product Details