The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198631.2 | NP_941033.2 | 10 - 11 | 1388 | 59 |
GenBank mRNA | AK129275.1 | - | 5 - 6 | 512 | 59 |
CF538051.1 | - | 6 - 7 | 602 | 59 |