Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc9
Entrez Gene ID
243846
Gene Name
coiled-coil domain containing 9
Gene Aliases
2600011L02Rik
Chromosome Location
Chr.7: 16273534 - 16286815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136471.1 NP_001129943.1 9 - 10 1083 57
NM_172297.1 NP_758501.2 8 - 9 1100 57
GenBank mRNA AK011186.1 - 2 - 3 1907 57
AK134892.1 - 9 - 10 1069 57
BC021915.1 - 8 - 9 1100 57

View Full Product Details